Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146067.1 | NP_001139539.1 | 1379 | 63 | ||
GenBank mRNA | BT063357.1 | - | 1379 | 63 | ||
EU974186.1 | - | 1331 | 63 | |||
FJ686697.1 | - | 1331 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Zea mays |