Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100283278 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001156180.1 | NP_001149652.1 | 556 | 94 | ||
GenBank mRNA | BT038967.1 | - | 500 | 94 | ||
EU963030.1 | - | 523 | 94 | |||
EU964129.1 | - | 556 | 94 |