Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sfp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111580.1 | NP_001105050.1 | 708 | 96 | ||
GenBank mRNA | AF355602.1 | - | 708 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Zea mays |