Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cl15444_1(721) | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001152502.1 | NP_001145974.1 | 444 | 99 | ||
GenBank mRNA | BT054355.1 | - | 444 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Zea mays |