Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112166.1 | NP_001105636.1 | 324 | 89 | ||
GenBank mRNA | AF305892.1 | - | 324 | 89 | ||
BT069123.1 | - | 249 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Zea mays |