Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ache | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112330.1 | NP_001105800.1 | 561 | 87 | ||
GenBank mRNA | AB093208.1 | - | 561 | 87 |