Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lac5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112451.1 | NP_001105921.1 | 1693 | 58 | ||
GenBank mRNA | AM086217.1 | - | 1693 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Zea mays |