Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100283583 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001156484.1 | NP_001149956.1 | 729 | 76 | ||
GenBank mRNA | EU965216.1 | - | 729 | 76 |