Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100281351 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001154269.1 | NP_001147741.1 | 399 | 77 | ||
GenBank mRNA | BT062039.1 | - | 360 | 77 | ||
EU956416.1 | - | 399 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Zea mays |