Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100283834 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001156732.1 | NP_001150204.1 | 1016 | 69 | ||
GenBank mRNA | EU966124.1 | - | 1016 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Kinase 3' Most Zea mays |