Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111417.1 | NP_001104887.1 | 383 | 86 | ||
GenBank mRNA | AF034944.1 | - | 383 | 86 | ||
BT035851.1 | - | 493 | 86 | |||
EU955122.1 | - | 370 | 86 | |||
EU955990.1 | - | 475 | 86 | |||
EU957882.1 | - | 378 | 86 | |||
EU958789.1 | - | 410 | 86 | |||
EU965717.1 | - | 538 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Zea mays |