Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ckb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111504.1 | NP_001104974.1 | 402 | 78 | ||
GenBank mRNA | AF239816.1 | - | 402 | 78 | ||
BT040828.1 | - | 377 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Zea mays |