Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111533.1 | NP_001105003.1 | 819 | 88 | ||
GenBank mRNA | AF271894.1 | - | 739 | 88 | ||
BT067411.1 | - | 777 | 88 | |||
DQ335760.1 | - | 819 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Zea mays |