Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111569.1 | NP_001105039.1 | 285 | 89 | ||
GenBank mRNA | AF330036.1 | - | 285 | 89 | ||
AF330037.1 | - | 136 | 89 | |||
BT040253.1 | - | 282 | 89 | |||
BT043171.1 | - | 320 | 89 | |||
EU961678.1 | - | 338 | 89 | |||
L39789.1 | - | 259 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Zea mays |