Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: frk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111740.1 | NP_001105210.1 | 744 | 92 | ||
GenBank mRNA | AY197772.1 | - | 744 | 92 | ||
BT034483.1 | - | 755 | 92 | |||
BT039818.1 | - | 800 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Zea mays |