Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112413.1 | NP_001105883.1 | 187 | 68 | ||
GenBank mRNA | AM075828.1 | - | 187 | 68 | ||
EU957714.1 | - | 270 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Zea mays |