Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112445.1 | NP_001105915.1 | 346 | 63 | ||
GenBank mRNA | AM086215.1 | - | 346 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Zea mays |