Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112551.1 | NP_001106021.1 | 1229 | 77 | ||
GenBank mRNA | EF515840.1 | - | 1229 | 77 |