Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112554.1 | NP_001106024.1 | 1188 | 54 | ||
GenBank mRNA | EF515844.1 | - | 1188 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Zea mays |