Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATTTCTCAAAAAAAAAATCGTTA[A/C]CAGCCATTTTGCCACTAATCTTACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609606 | ||||||||||||||||||||
Literature Links: |
ATG3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATG3 - autophagy related 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278712.1 | 1339 | Missense Mutation | GTA,TTA | V,L 302 | NP_001265641.1 | |
NM_022488.4 | 1339 | Intron | NP_071933.2 | |||
XM_011513074.1 | 1339 | Intron | XP_011511376.1 |