Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATTCTCAGTCCTTATCCCACTGA[C/T]CTCTCAGCAGCATTTGCCGTAGTAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 606987 | |||||||||||||||||||||||
Literature Links: |
GRK7 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
GRK7 - G protein-coupled receptor kinase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139209.2 | 1331 | Intron | NP_631948.1 | |||
XM_017005717.1 | 1331 | UTR 5 | XP_016861206.1 |