Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACCCAGCCAGGGCCCTGCAGCAA[A/C]TGAGGTAATGAGCCTTAGAATAAAC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 606639 | |||||||||||||||||||||||
Literature Links: |
GFM1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
GFM1 - G elongation factor mitochondrial 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308164.1 | 706 | Missense Mutation | ATG,CTG | M,L 190 | NP_001295093.1 | |
NM_001308166.1 | 706 | Missense Mutation | ATG,CTG | M,L 190 | NP_001295095.1 | |
NM_024996.5 | 706 | Missense Mutation | ATG,CTG | M,L 190 | NP_079272.4 | |
XM_006713795.1 | 706 | Missense Mutation | ATG,CTG | M,L 190 | XP_006713858.1 |