Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAACCTAAACACACGCACTTGAC[A/G]GGGGTATGCCAAAGGGGCACACGTC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
ZNF639 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ZNF639 - zinc finger protein 639 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303425.1 | Intron | NP_001290354.1 | ||||
NM_001303426.1 | Intron | NP_001290355.1 | ||||
NM_016331.2 | Intron | NP_057415.1 | ||||
XM_017006550.1 | Intron | XP_016862039.1 | ||||
XM_017006551.1 | Intron | XP_016862040.1 | ||||
XM_017006552.1 | Intron | XP_016862041.1 | ||||
XM_017006553.1 | Intron | XP_016862042.1 |