Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTGATTAAGGCAGAGCCTGTGCA[A/G]GAGGGAAGGAGACCTAGACTCCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616238 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYAR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LYAR - Ly1 antibody reactive | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZBTB49 - zinc finger and BTB domain containing 49 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145291.3 | Intron | NP_660334.3 | ||||
XM_005247951.4 | Intron | XP_005248008.1 | ||||
XM_006713864.3 | Intron | XP_006713927.1 | ||||
XM_011513416.1 | Intron | XP_011511718.1 | ||||
XM_017007835.1 | Intron | XP_016863324.1 | ||||
XM_017007836.1 | Intron | XP_016863325.1 |