Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTTTGCAGGAGGTCTACTGACCC[C/T]TCCATTCGAGTAACCTTCTCTTCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 123825 | ||||||||||||||||||||
Literature Links: |
CNGA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CNGA1 - cyclic nucleotide gated channel alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000087.3 | 2214 | Silent Mutation | GAA,GAG | E,E 631 | NP_000078.2 | |
NM_001142564.1 | 2214 | Silent Mutation | GAA,GAG | E,E 700 | NP_001136036.1 | |
XM_005248049.4 | 2214 | Silent Mutation | GAA,GAG | E,E 706 | XP_005248106.2 | |
XM_011513623.2 | 2214 | Silent Mutation | GAA,GAG | E,E 631 | XP_011511925.1 | |
XM_017007712.1 | 2214 | Silent Mutation | GAA,GAG | E,E 631 | XP_016863201.1 |
LOC101927157 - uncharacterized LOC101927157 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |