Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCCAATCCGTGTGCCTTTTATTT[A/C]TTTTTCTTGACTTACTGTACTGGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TBC1D19 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TBC1D19 - TBC1 domain family member 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001292054.1 | Intron | NP_001278983.1 | ||||
NM_018317.3 | Intron | NP_060787.2 | ||||
XM_011513852.2 | Intron | XP_011512154.1 | ||||
XM_017008363.1 | Intron | XP_016863852.1 | ||||
XM_017008364.1 | Intron | XP_016863853.1 |