Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGTTTCTCCCTCCCTCTGTTCCT[A/C]TCTCCCTCCCTCGGGTTTGCGTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610697 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDZD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDZD2 - PDZ domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178140.3 | Intron | NP_835260.2 | ||||
XM_005248269.4 | Intron | XP_005248326.1 | ||||
XM_005248271.1 | Intron | XP_005248328.1 | ||||
XM_005248272.3 | Intron | XP_005248329.1 | ||||
XM_006714460.2 | Intron | XP_006714523.1 | ||||
XM_011513992.2 | Intron | XP_011512294.1 | ||||
XM_011513993.2 | Intron | XP_011512295.1 | ||||
XM_011513994.2 | Intron | XP_011512296.1 | ||||
XM_011513995.2 | Intron | XP_011512297.1 | ||||
XM_011513996.2 | Intron | XP_011512298.1 | ||||
XM_017009245.1 | Intron | XP_016864734.1 | ||||
XM_017009246.1 | Intron | XP_016864735.1 |