Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAATAACGTGGTCCATTTTGTACA[C/T]GGAGAAATTTGGCCTAGTTTAGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606184 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTS12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAMTS12 - ADAM metallopeptidase with thrombospondin type 1 motif 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324511.1 | 7460 | Intron | NP_001311440.1 | |||
NM_001324512.1 | 7460 | UTR 3 | NP_001311441.1 | |||
NM_030955.3 | 7460 | UTR 3 | NP_112217.2 | |||
XM_017009905.1 | 7460 | UTR 3 | XP_016865394.1 | |||
XM_017009906.1 | 7460 | UTR 3 | XP_016865395.1 | |||
XM_017009907.1 | 7460 | UTR 3 | XP_016865396.1 | |||
XM_017009908.1 | 7460 | UTR 3 | XP_016865397.1 | |||
XM_017009909.1 | 7460 | UTR 3 | XP_016865398.1 |