Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGATGTCCCAGGCCCCACCTGACA[A/G]TTCTTGTAGTCTCACAGGAGATGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606586 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105374753 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105374753 - uncharacterized LOC105374753 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAI14 - retinoic acid induced 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145520.1 | Intron | NP_001138992.1 | ||||
NM_001145521.1 | Intron | NP_001138993.1 | ||||
NM_001145522.1 | Intron | NP_001138994.1 | ||||
NM_001145523.1 | Intron | NP_001138995.1 | ||||
NM_001145525.1 | Intron | NP_001138997.1 | ||||
NM_015577.2 | Intron | NP_056392.2 | ||||
XM_006714469.2 | Intron | XP_006714532.1 | ||||
XM_011514016.2 | Intron | XP_011512318.2 | ||||
XM_011514017.2 | Intron | XP_011512319.1 | ||||
XM_011514018.1 | Intron | XP_011512320.1 | ||||
XM_011514019.1 | Intron | XP_011512321.1 | ||||
XM_011514020.1 | Intron | XP_011512322.1 | ||||
XM_011514021.1 | Intron | XP_011512323.1 | ||||
XM_011514022.1 | Intron | XP_011512324.1 | ||||
XM_011514023.1 | Intron | XP_011512325.1 | ||||
XM_011514024.1 | Intron | XP_011512326.1 | ||||
XM_011514025.2 | Intron | XP_011512327.2 | ||||
XM_017009334.1 | Intron | XP_016864823.1 | ||||
XM_017009335.1 | Intron | XP_016864824.1 | ||||
XM_017009336.1 | Intron | XP_016864825.1 |