Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGATTTTACCCCAGTGAAGTAAA[A/C]CCCAGCGGCCGCCATCTCCTGTGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600355 | ||||||||||||||||||||
Literature Links: |
NAIP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NAIP - NLR family apoptosis inhibitory protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004536.2 | 974 | Missense Mutation | GTT,TTT | V,F 86 | NP_004527.2 | |
NM_022892.1 | 974 | Intron | NP_075043.1 | |||
XM_011543413.2 | 974 | Intron | XP_011541715.1 | |||
XM_017009490.1 | 974 | Intron | XP_016864979.1 |