Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTCTCTAAATGTGACAAATATAA[C/T]AAAGTGAGTCTAAAAAAATAAAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604798 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HOMER1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HOMER1 - homer scaffolding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277077.1 | Intron | NP_001264006.1 | ||||
NM_001277078.1 | Intron | NP_001264007.1 | ||||
NM_004272.4 | Intron | NP_004263.1 | ||||
XM_017010059.1 | Intron | XP_016865548.1 |