Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCTCTAATTTCTGAGCCTTTGCA[A/G]GGCAAATTAGTTCATTCTCATTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614667 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTO1 - mitochondrial tRNA translation optimization 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001123226.1 | Intron | NP_001116698.1 | ||||
NM_012123.3 | Intron | NP_036255.2 | ||||
NM_133645.2 | Intron | NP_598400.1 | ||||
XM_006715444.2 | Intron | XP_006715507.1 | ||||
XM_006715445.2 | Intron | XP_006715508.1 | ||||
XM_006715446.3 | Intron | XP_006715509.1 | ||||
XM_011535723.1 | Intron | XP_011534025.1 | ||||
XM_011535724.1 | Intron | XP_011534026.1 | ||||
XM_011535725.2 | Intron | XP_011534027.1 | ||||
XM_017010697.1 | Intron | XP_016866186.1 | ||||
XM_017010698.1 | Intron | XP_016866187.1 | ||||
XM_017010699.1 | Intron | XP_016866188.1 | ||||
XM_017010700.1 | Intron | XP_016866189.1 |