Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTAGTTTAATTAAGCAAAGGTAT[C/T]AGGAGGTCTGTTTCAGCTCATTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604322 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC17A5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC17A5 - solute carrier family 17 member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012434.4 | 1742 | UTR 3 | NP_036566.1 | |||
XM_005248710.3 | 1742 | UTR 3 | XP_005248767.1 | |||
XM_005248711.2 | 1742 | UTR 3 | XP_005248768.1 | |||
XM_011535750.2 | 1742 | UTR 3 | XP_011534052.1 | |||
XM_017010730.1 | 1742 | UTR 3 | XP_016866219.1 | |||
XM_017010731.1 | 1742 | UTR 3 | XP_016866220.1 |