Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCTTCTGATCCATTTATCTCCAG[C/T]AACATTAGCATAGCATAGGTAATAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 172100 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PGM3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PGM3 - phosphoglucomutase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RWDD2A - RWD domain containing 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322335.1 | 659 | Intron | NP_001309264.1 | |||
NM_001322336.1 | 659 | Intron | NP_001309265.1 | |||
NM_001322337.1 | 659 | Intron | NP_001309266.1 | |||
NM_001322339.1 | 659 | Intron | NP_001309268.1 | |||
NM_033411.4 | 659 | Intron | NP_219479.2 | |||
XM_017010222.1 | 659 | UTR 5 | XP_016865711.1 | |||
XM_017010223.1 | 659 | UTR 5 | XP_016865712.1 |