Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCTGAATTCATTTTTTCTTATTC[C/T]TAGTCTTCTGTATTTTAACATTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611470 MIM: 601585 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LGSN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LGSN - lengsin, lens protein with glutamine synthetase domain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143940.1 | Intron | NP_001137412.1 | ||||
NM_016571.2 | Intron | NP_057655.2 | ||||
XM_011535889.2 | Intron | XP_011534191.1 | ||||
XM_011535892.2 | Intron | XP_011534194.1 | ||||
XM_017010929.1 | Intron | XP_016866418.1 | ||||
XM_017010930.1 | Intron | XP_016866419.1 | ||||
XM_017010931.1 | Intron | XP_016866420.1 |
PTP4A1 - protein tyrosine phosphatase type IVA, member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003463.4 | Intron | NP_003454.1 | ||||
XM_011536111.1 | Intron | XP_011534413.1 | ||||
XM_011536112.1 | Intron | XP_011534414.1 | ||||
XM_017011270.1 | Intron | XP_016866759.1 | ||||
XM_017011271.1 | Intron | XP_016866760.1 | ||||
XM_017011272.1 | Intron | XP_016866761.1 |