Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAAACCAGTTTAGTTTCATCAGG[A/G]ATTTTTTTTTTTTTGCATCAATTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194540 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIVEP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HIVEP1 - human immunodeficiency virus type I enhancer binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002114.3 | Intron | NP_002105.3 | ||||
XM_011514546.2 | Intron | XP_011512848.1 | ||||
XM_011514547.2 | Intron | XP_011512849.1 | ||||
XM_011514548.2 | Intron | XP_011512850.1 | ||||
XM_011514549.2 | Intron | XP_011512851.1 | ||||
XM_011514550.2 | Intron | XP_011512852.1 | ||||
XM_011514551.2 | Intron | XP_011512853.1 | ||||
XM_011514552.2 | Intron | XP_011512854.1 | ||||
XM_011514553.1 | Intron | XP_011512855.1 | ||||
XM_011514555.2 | Intron | XP_011512857.1 | ||||
XM_017010800.1 | Intron | XP_016866289.1 | ||||
XM_017010801.1 | Intron | XP_016866290.1 | ||||
XM_017010802.1 | Intron | XP_016866291.1 | ||||
XM_017010803.1 | Intron | XP_016866292.1 | ||||
XM_017010804.1 | Intron | XP_016866293.1 |
LOC101928253 - uncharacterized LOC101928253 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |