Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAGGGTTTATAGACTTAGCTCTC[C/T]TCTCTGGATGAGATACTGTCAGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613594 MIM: 613595 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTN3A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BTN3A3 - butyrophilin subfamily 3 member A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242803.1 | Intron | NP_001229732.1 | ||||
NM_006994.4 | Intron | NP_008925.1 | ||||
NM_197974.2 | Intron | NP_932078.2 |