Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCATCCTGTGTCACCTAAAACTCC[A/G]CTTAGGAAGATAAGAGGCATTCATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606993 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IP6K3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IP6K3 - inositol hexakisphosphate kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142883.1 | 2152 | UTR 3 | NP_001136355.1 | |||
NM_054111.4 | 2152 | UTR 3 | NP_473452.2 | |||
XM_005248842.3 | 2152 | UTR 3 | XP_005248899.1 | |||
XM_005248843.3 | 2152 | UTR 3 | XP_005248900.1 | |||
XM_011514295.2 | 2152 | Intron | XP_011512597.1 |