Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTAACCATATCTTAGTTGGGCTT[C/T]TTAGAGTAGATGTGTTCACATTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602338 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM217A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM217A - family with sequence similarity 217 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173563.2 | 2196 | UTR 3 | NP_775834.2 | |||
XM_005248951.3 | 2196 | UTR 3 | XP_005249008.1 | |||
XM_006715025.2 | 2196 | UTR 3 | XP_006715088.1 | |||
XM_006715026.3 | 2196 | UTR 3 | XP_006715089.1 | |||
XM_011514413.2 | 2196 | UTR 3 | XP_011512715.1 | |||
XM_011514414.2 | 2196 | UTR 3 | XP_011512716.1 | |||
XM_011514415.1 | 2196 | UTR 3 | XP_011512717.1 | |||
XM_011514416.2 | 2196 | UTR 3 | XP_011512718.1 | |||
XM_011514417.1 | 2196 | UTR 3 | XP_011512719.1 | |||
XM_017010480.1 | 2196 | UTR 3 | XP_016865969.1 | |||
XM_017010481.1 | 2196 | UTR 3 | XP_016865970.1 | |||
XM_017010482.1 | 2196 | UTR 3 | XP_016865971.1 |
PRPF4B - pre-mRNA processing factor 4B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |