Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGCCTTTCATAAGAGATACACA[C/T]TTCTACTTTGTTTTGGGATACCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611805 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELOVL5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ELOVL5 - ELOVL fatty acid elongase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242828.1 | Intron | NP_001229757.1 | ||||
NM_001242830.1 | Intron | NP_001229759.1 | ||||
NM_001242831.1 | Intron | NP_001229760.1 | ||||
NM_001301856.1 | Intron | NP_001288785.1 | ||||
NM_021814.4 | Intron | NP_068586.1 |
RPS16P5 - ribosomal protein S16 pseudogene 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |