Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGCTGTCCCGACCTTTGACATCG[C/T]GCCCCAGCTGGGGTCTGTGATGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601970 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC00689 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC00689 - long intergenic non-protein coding RNA 689 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VIPR2 - vasoactive intestinal peptide receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304522.1 | 3050 | UTR 3 | NP_001291451.1 | |||
NM_001308259.1 | 3050 | UTR 3 | NP_001295188.1 | |||
NM_003382.4 | 3050 | UTR 3 | NP_003373.2 | |||
XM_005249561.3 | 3050 | UTR 3 | XP_005249618.1 | |||
XM_006716107.2 | 3050 | Intron | XP_006716170.1 | |||
XM_006716108.3 | 3050 | Intron | XP_006716171.1 | |||
XM_011516550.2 | 3050 | Intron | XP_011514852.1 | |||
XM_017012580.1 | 3050 | UTR 3 | XP_016868069.1 |