Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGAGGTCTATTCAATTTCTGTGA[A/G]AAGGAAGACGTGAATTTACCTTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612249 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
THSD7A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
THSD7A - thrombospondin type 1 domain containing 7A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015204.2 | 8730 | UTR 3 | NP_056019.1 | |||
XM_006715659.1 | 8730 | UTR 3 | XP_006715722.1 | |||
XM_006715660.1 | 8730 | UTR 3 | XP_006715723.1 | |||
XM_006715662.1 | 8730 | UTR 3 | XP_006715725.1 | |||
XM_011515193.2 | 8730 | UTR 3 | XP_011513495.1 | |||
XM_011515194.1 | 8730 | UTR 3 | XP_011513496.1 |