Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATAAAAACTCAACCAAAAAGTGG[A/C]ATCAAAACAGAACTTAAAAAATTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600541 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ETV1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ETV1 - ETS variant 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163147.1 | 4177 | UTR 3 | NP_001156619.1 | |||
NM_001163148.1 | 4177 | UTR 3 | NP_001156620.1 | |||
NM_001163149.1 | 4177 | UTR 3 | NP_001156621.1 | |||
NM_001163150.1 | 4177 | UTR 3 | NP_001156622.1 | |||
NM_001163151.1 | 4177 | UTR 3 | NP_001156623.1 | |||
NM_001163152.1 | 4177 | UTR 3 | NP_001156624.1 | |||
NM_004956.4 | 4177 | UTR 3 | NP_004947.2 | |||
XM_005249635.4 | 4177 | UTR 3 | XP_005249692.1 | |||
XM_005249636.4 | 4177 | UTR 3 | XP_005249693.1 | |||
XM_011515167.2 | 4177 | UTR 3 | XP_011513469.1 | |||
XM_011515168.2 | 4177 | UTR 3 | XP_011513470.1 | |||
XM_011515170.2 | 4177 | UTR 3 | XP_011513472.1 |