Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCATGTATAATCTCTCTGTGAAAA[C/T]AGGTGTGTAGGTGTTTGCCCAGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611119 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHL7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLHL7 - kelch like family member 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUPL2 - nucleoporin like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007342.2 | Intron | NP_031368.1 | ||||
XM_005249592.1 | Intron | XP_005249649.1 | ||||
XM_005249593.3 | Intron | XP_005249650.1 | ||||
XM_011515104.2 | Intron | XP_011513406.1 | ||||
XM_017011714.1 | Intron | XP_016867203.1 | ||||
XM_017011715.1 | Intron | XP_016867204.1 | ||||
XM_017011716.1 | Intron | XP_016867205.1 | ||||
XM_017011717.1 | Intron | XP_016867206.1 | ||||
XM_017011718.1 | Intron | XP_016867207.1 |