Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTTGGTGTGACAATGAAAAATCA[C/T]GCATTCTTTTCCTGTAATTATTTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606732 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OSBPL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OSBPL3 - oxysterol binding protein like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015550.3 | 4537 | UTR 3 | NP_056365.1 | |||
NM_145320.2 | 4537 | UTR 3 | NP_663160.1 | |||
NM_145321.2 | 4537 | UTR 3 | NP_663161.1 | |||
NM_145322.2 | 4537 | UTR 3 | NP_663162.1 | |||
XM_005249698.3 | 4537 | UTR 3 | XP_005249755.1 | |||
XM_006715681.3 | 4537 | UTR 3 | XP_006715744.1 | |||
XM_006715682.3 | 4537 | UTR 3 | XP_006715745.1 | |||
XM_006715683.3 | 4537 | UTR 3 | XP_006715746.1 | |||
XM_006715684.3 | 4537 | UTR 3 | XP_006715747.1 | |||
XM_011515258.2 | 4537 | UTR 3 | XP_011513560.1 | |||
XM_011515259.2 | 4537 | UTR 3 | XP_011513561.1 | |||
XM_017011948.1 | 4537 | UTR 3 | XP_016867437.1 | |||
XM_017011949.1 | 4537 | UTR 3 | XP_016867438.1 |