Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAACAGCTACTACTATTCTATGC[C/T]ATGGCTCTATCTTTTTTTTAAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610555 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1GALT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1GALT1 - core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020156.4 | Intron | NP_064541.1 | ||||
XM_005249812.1 | Intron | XP_005249869.1 | ||||
XM_011515453.2 | Intron | XP_011513755.1 | ||||
XM_011515455.2 | Intron | XP_011513757.1 | ||||
XM_011515456.2 | Intron | XP_011513758.1 | ||||
XM_017012442.1 | Intron | XP_016867931.1 | ||||
XM_017012443.1 | Intron | XP_016867932.1 | ||||
XM_017012444.1 | Intron | XP_016867933.1 | ||||
XM_017012445.1 | Intron | XP_016867934.1 | ||||
XM_017012446.1 | Intron | XP_016867935.1 | ||||
XM_017012447.1 | Intron | XP_016867936.1 | ||||
XM_017012448.1 | Intron | XP_016867937.1 | ||||
XM_017012449.1 | Intron | XP_016867938.1 |