Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTAAACCATTTTCGTGTCTTGC[A/G]TCTCCTGCTATTCTTGGTGAGTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611019 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTR3C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACTR3C - ARP3 actin-related protein 3 homolog C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ATP6V0E2 - ATPase H+ transporting V0 subunit e2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100592.2 | Intron | NP_001094062.1 | ||||
NM_001289990.1 | Intron | NP_001276919.1 | ||||
NM_145230.3 | Intron | NP_660265.2 |
ATP6V0E2-AS1 - ATP6V0E2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF862 - zinc finger protein 862 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |