Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACAGAGCCCGTGCACACAGTTCT[A/G]TGGTCGCACCTAGCTGTGGGAGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTR3B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACTR3B - ARP3 actin related protein 3 homolog B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040135.2 | 6280 | Intron | NP_001035225.1 | |||
NM_020445.5 | 6280 | Intron | NP_065178.1 | |||
XM_006716065.2 | 6280 | Intron | XP_006716128.1 | |||
XM_006716066.1 | 6280 | Intron | XP_006716129.1 | |||
XM_006716067.3 | 6280 | UTR 3 | XP_006716130.1 | |||
XM_011516425.2 | 6280 | Intron | XP_011514727.1 | |||
XM_011516426.2 | 6280 | Intron | XP_011514728.1 | |||
XM_011516427.1 | 6280 | Intron | XP_011514729.1 | |||
XM_017012460.1 | 6280 | Intron | XP_016867949.1 | |||
XM_017012461.1 | 6280 | Intron | XP_016867950.1 | |||
XM_017012462.1 | 6280 | Intron | XP_016867951.1 | |||
XM_017012463.1 | 6280 | Intron | XP_016867952.1 | |||
XM_017012464.1 | 6280 | Intron | XP_016867953.1 | |||
XM_017012465.1 | 6280 | Intron | XP_016867954.1 |