Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGTAGCTTCTTTGTATTAATCTTT[C/T]TTGGGTCACAGTGCCTTGACAGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603431 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF4H PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EIF4H - eukaryotic translation initiation factor 4H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022170.1 | Intron | NP_071496.1 | ||||
NM_031992.1 | Intron | NP_114381.1 | ||||
XM_011516556.1 | Intron | XP_011514858.1 | ||||
XM_011516557.1 | Intron | XP_011514859.1 |