Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCTACCCTAAGTAGCTGGAACTA[C/T]AGGCATGAGCTACTGCATCAGGCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616714 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HBP1 - HMG-box transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244262.1 | Intron | NP_001231191.1 | ||||
NM_012257.3 | Intron | NP_036389.2 | ||||
XM_005250266.2 | Intron | XP_005250323.1 | ||||
XM_005250267.2 | Intron | XP_005250324.1 | ||||
XM_017011967.1 | Intron | XP_016867456.1 |